Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902756

Sigma-Aldrich

Imidazole-1-sulfonyl azide, HCl salt

Synonym(s):

1H-Imidazole-1-sulfonyl azide, HCl salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H3N5O2S · HCl
CAS Number:
Molecular Weight:
209.61
MDL number:
UNSPSC Code:
12352200

assay

≥95%

form

powder

reaction suitability

reaction type: click chemistry

mp

100-105 °C

storage temp.

2-8°C

SMILES string

Cl.[N-]=[N+]=NS(=O)(=O)n1ccnc1

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service