Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902616

Sigma-Aldrich

(4-Methoxybenzylidene)aniline nitrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO2
CAS Number:
Molecular Weight:
227.26
UNSPSC Code:
12352005
NACRES:
NA.22

assay

≥95%

form

solid

mp

119 °C

storage temp.

2-8°C

InChI

1S/C14H13NO2/c1-17-14-9-7-12(8-10-14)11-15(16)13-5-3-2-4-6-13/h2-11H,1H3

InChI key

GIBHBSRKHBYUKU-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service