Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

902470

Sigma-Aldrich

Activated carbon

Bis(2-ethylhexyl)phosphate modified

Synonym(s):

Activated charcoal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101522
NACRES:
NA.23

form

powder

Looking for similar products? Visit Product Comparison Guide

Application

Activated carbons are internally porous microcrystalline, non-graphitic forms of carbon. The activated carbons possess large surface area (∼1000 m2/g) and pore volumes which make them useful for applications in energy storage devices, catalysis, and for removal of impurities from gases and liquids[1][2].Our activated carbon has been modified with Bis(2-ethylhexyl)phosphate and is well suited for metal ion removal from water.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Energy storage applications of activated carbons: supercapacitors and hydrogen storage.
Sevilla M, et al.
Energy & Environmental Science, 7, 1250-1280 (2014)
Activated Carbon for Water and Wastewater Treatment: Integration of Adsorption and Biological Treatment
Cecen F, et al.
Wiley-VCH (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service