Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902209

Sigma-Aldrich

Cesium dodecahydroxydodecaborate

Synonym(s):

Boric acid (H14B12O12), dicesium salt, Dicesium dodecahydroxy-closo-dodecaborate, Dicesium dodecahydroxydodecaborate(2-), Dodecaborate(2-), dodecahydroxy-, dicesium (9CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B12H12O12·2Cs
CAS Number:
Molecular Weight:
599.63
UNSPSC Code:
12352302
NACRES:
NA.23
form:
powder

form

powder

storage temp.

2-8°C

Application

Cesium dodecahydroxydodecaborate, a dicesium salt of the icosahedral borane anion dodecahydroxy-closo-dodecarborate(2-) (closo-Cs2B12(OH)22), is potentially used in drug delivery systems, radio-imaging reagents and hydrophobic space-filling pharmacophores.[1][2]
Used as a precursor in SN2 reactions of alkyl or benzyl halides to form derivatives such as Dodeca(2,3,4,5,6-pentafluorobenzyloxy)-hypercloso-dodecaborane.

Legal Information

Sold under Material Transfer Agreement with Prof. Alexander Spokoyny from Department of Chemistry and Biochemistry, UCLA.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Study of the Sequential Acid-Catalyzed Hydroxylation of Dodecahydro-c loso-dodecaborate (2-)
Peymann T, et al.
Inorganic Chemistry, 39(6), 1163-1170 (2000)
An Improved Method for the Synthesis of [closo-B12 (OH) 12]-2
Bayer MJ and Hawthorne MF
Inorganic Chemistry, 43(6), 2018-2020 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service