Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901969

Sigma-Aldrich

Carbon/graphite screen printing paste

fast drying, suitable for screen printed working electrodes

Synonym(s):

C2000802P2, conductive ink, graphite/carbon ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
UNSPSC Code:
12352200
NACRES:
NA.23
Product 901969 is not currently sold in your country. Contact Technical Service

description

Coverage: 305 cm2/g (using a 230 mesh stainless steel screen)
Cured Thickness: 15 micron (printed through 230 stainless steel mesh with 13 micron emulsion)
Sheet resistivity: < 50Ω/square at 25 μm (printed through 230 stainless steel mesh with 13 micron emulsion)

form

paste

composition

Solid content, 39-41% (at 130°C)

viscosity

2.0-3.5 Pa.s (Haake RS1 C20/2º TiL at 230 sec-1 at 25 °C.)

Looking for similar products? Visit Product Comparison Guide

General description

Carbon/graphite screen printing paste is a carbon containing ink that can be used in the formation of working and counter electrodes. The electrode formed can be modified using an ionophore to obtain good cathodic peaks and cyclic voltammetric values. These electrodes can further be used in the fabrication of electrochemical devices.[1][2]
This product is designed to be used for screen-printing working electrodes. This ink has been optimised to give superior electrochemical performance. It combines good line definition and good electrochemical performance with good reversibility when using cyclic voltammetry.

Application

Carbon/graphite screen printing paste can be coated on flexible substrates for the formation of electrochemical sensors for biological detection.[1][2][3][4]

Features and Benefits

This ink can be used in conjunction with UV dielectrics and ink is fast drying.

Preparation Note

PROCESSING:
PRODUCT MUST BE STIRRED PRIOR TO USE.
Screen Printing Equipment: Semi-Automatic, manual
Ink Screen Life: >3 hours
Screen Types: Stainless steel, polyester, mesh 156-230 tpi

Typical Drying Conditions :
For box-oven drying, 60 °C for 30 minutes or 80 °C for 10-15 minutes
For belt-drier ovens, 130 °C for 10 minutes of element exposure
Clean-up solvent : ethoxy propanol or sericol
Substrate: polyester, PVC, polycarbonate or ceramic.

Legal Information

Product of Gwent Electronic Materials Ltd (GEM), part of Sun Chemical Corporation.

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

114.8 °F - closed cup

flash_point_c

46 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical determination of antihypertensive drugs by employing costless and portable unmodified screen-printed electrodes
Khorshed AA, et al.
Talanta, 198(6), 447-456 (2019)
Pseudo Cavity of Schiff Base Ionophore Incorporated in Screen Printed Electrode for Sensing of Zn (II)
Rana S, et al.
Journal of the Electrochemical Society, 166(6), B464-B471 (2019)
Hybrid electrochemical sensor platform for capsaicin determination using coarsely stepped cyclic squarewave voltammetry
Sopstad S, et al.
Biosensors And Bioelectronics, 130(6), 374-381 (2019)
Forensic electrochemistry: simultaneous voltammetric detection of MDMA and its fatal counterpart "Dr Death" (PMA)
Cumba L R, et al.
Analytical Methods : Advancing Methods and Applications, 8(1), 142-152 (2016)
Highly sensitive screen printed strain sensors on flexible substrates via ink composition optimization
Anderson N, et al.
Sensors and actuators A, Physical, 290(6), 1-7 (2019)

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service