Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901876

Sigma-Aldrich

SunTronic®aqueous silver flexo ink

Synonym(s):

Ag ink, Conductive ink, Silver ink, SunTronic®AFT6700

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Sheet resistivity: 15 mΩ/sq/mil
Specific gravity: 2.01g/cm3
VOC: <1.2 wt%
Volume Resistivity: <37.5 Ω.cm

form

liquid

viscosity

19-21 poise

Storage temp.

2-8°C

General description

SunTronic aqueous silver flexo ink AFT6700 is a water-based silver conductive ink designed for the production of printed antenna for radio frequency identification (RFID), conductive interconnects, and printed circuitry via flexographic printing.
AFT6700 is appropriate for paper and cardboard substrates and also water-receptive polymer films and provides a unique combination of high-speed printability, fast drying, and conductivity in an easy-to-use ink system. AFT6700 can also be printed on plastic films bearing a water-receptive primer.

Caution

Do not freeze.

Preparation Note

Processing and Handling Guidelines
PRINTING


AFT6700 is supplied as ready-for-use ink. Thinning is not recommended. Soft sediment may occur over time so thorough mixing prior to use is necessary.
The below details are only a guide, and the final choice of flexo printing specifications may depend on substrate and web speed. To achieve maximum coverage, high volume anilox rolls are recommended (80-120 lpi with 14-22 BCM). As an example, thickness of 3-4 microns can be achieved with a 160 line/inch and 20 BCM ART anilox (Praxair Technology, Inc.). Print tests have demonstrated that a soft flexographic plate such as Nyloflex ART (BASF) produces the highest quality graphics when used in conjunction with a medium-hard sticky back. A closed doctor-blade chamber is recommended.

Continuous anilox rotation is required throughout the print run to avoid ink drying on flexo plate and anilox roll. AFT6700 should not be allowed to dry completely on printing equipment. Prompt cleaning with water is highly recommended. Anilox rolls should be also cleaned using a sonicating bath after each print run.

DRYING


AFT6700 is a fast drying ink that will dry to the touch between 150-300 feet per minute using standard dryers found on flexographic presses. Maximum conductivity can be achieved with further drying, inline or offline, using infrared (IR) dryers, high capacity forced hot air ovens and/or heated mandrels.

Optimum drying conditions should be established for particular equipment used by the customer.

Legal Information

SunTronic is a registered trademark of Sun Chemical, Inc.

Pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service