Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901630

Sigma-Aldrich

Electroless nickel plating strike

Synonym(s):

Electroless nickel plating solution for copper, brass and copper alloys

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

form

liquid

color

green

optimum pH

3.2

General description

An electroless nickel plating process designed for electroless nickel plating on copper, brass and copper alloy
CLEANING PROCEDURE FOR COPPER SAMPLES:
1. Clean copper with some physical abrasion.
2. Dip for one minute in alkaline cleaner at 65°C (150°F).
3. Rinse with deionized water.
4. Dip for one minute in 30% HCl
5. Rinse with deionized water.
PLATING BATH - USE AND CONTROL
Operate bath at 95 °C. Plate for approximately one minute until surface of sample is completely nickel plated, then rinse with water. For build up of nickel deposit, use Nickelex (electroless nickel plating solution, ammonia free). Maintain pH of plating solution at 2.8 to 3.5 by adding ammonium hydroxide when pH falls below 2.8.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

230.7 °F

flash_point_c

110.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service