Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901463

Sigma-Aldrich

4,7-Bis(5-bromo-2-thienyl)-5,6-bis(dodecyloxy)-2,1,3-benzothiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H54Br2N2O2S3
CAS Number:
Molecular Weight:
826.85
UNSPSC Code:
12352101
NACRES:
NA.23

assay

≥97%

form

powder

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jicheng Zhang et al.
Chemical communications (Cambridge, England), 52(3), 469-472 (2015-11-10)
A new 1,8-naphthalimide based planar small molecular acceptor and two benzothiadiazole based wide band gap (WBG) polymer donors P1 and P2 were synthesized for nonfullerene organic photovoltaic cells (OPVs). Devices based on fluorinated polymer P2 achieved a highly improved PCE
A 1,8-naphthalimide based small molecular acceptor for polymer solar cells with high open circuit voltage.
Zhang X, et al.
Journal of Material Chemistry C, 3(27), 6979-6985 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service