Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901426

Sigma-Aldrich

Propargyl methacrylate

≥98.0%, contains 200 ppm MEHQ as inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12162002

assay

≥98.0%

form

liquid

color

colorless to pale yellow

storage temp.

−20°C

InChI

1S/C7H8O2/c1-4-5-9-7(8)6(2)3/h1H,2,5H2,3H3

InChI key

PZAWASVJOPLHCJ-UHFFFAOYSA-N

General description

Propargyl methacrylate is a methacrylate-based monomer for use in the synthesis of alkyne-functionalized polymers and copolymers. The incorporation of alkynes allows for rapid post-polymerization modification through reactions, such as copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) or thiol-yne click reactions. These reactions are highly preferable for functionalization due their high efficiency and the ability to easily install the corresponding functionality (e.g., azides or thiols) onto the biomolecule or small molecule of interest.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

120.2 °F

flash_point_c

49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembling amphiphilic poly(propargyl methacrylate) grafted DNA copolymers into multi-strand helices
Meng Y F, et al.
Soft Matter, 11, 5610- 513 (2015)
Clickable and Antifouling Platform of Poly[(propargyl methacrylate)-ran-(2-methacryloyloxyethyl phosphorylcholine)] for Biosensing Applications
Wiarachai O, et al.
Langmuir, 32 (4), 1184-1194 (2016)
Postfunctionalization of poly(propargyl methacrylate) using copper catalyzed 1,3-dipolar Huisgen cycloaddition: An easy route to electro-optic materials
Scarpaci A. et al.
Journal of Polymer Science: Part A, General Papers, 47, 5652- 560 (2009)
Jincai Wang et al.
Journal of chromatography. A, 1628, 461481-461481 (2020-08-22)
A single-step copolymerization strategy was developed for the preparation of carbohydrate (glucose and maltose) functionalized monoliths using click reaction. Firstly, novel carbohydrate-functionalized methacrylate monomers were synthesized through Cu(I)-catalyzed 1,3-dipolar cycloaddition (alkyne-azide reaction) of terminal alkyne with azide of carbohydrate derivatives.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service