901385
Poly(ethylene glycol) linear dendron
NHBoc terminated, PEG Mn 5000, generation 4
Synonym(s):
Dendron functionalized PEG, PFD-G4-PEG5k-NHBOC
About This Item
form
solid
mol wt
PEG Mn 5000
color
white to off-white
storage temp.
−20°C
SMILES string
CC(COC(CCNC(OC(C)(C)C)=O)=O)(C(OCC(C(OCC(C(OCC(C(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC
General description
- Polymer architecture: PEG-linear dendron
- Generation: 4
- PEG Average Mn: 5,000
- End Group Functionality: Boc-protected amine
- No. Surface Groups: 16
- Calculated Mol. Wt.: 9474.91 g/mol
- Boc : tert-Butyloxycarbonyl
- Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service