Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901339

Sigma-Aldrich

DCBPy

Synonym(s):

(2,5-Di-9H-carbazol-9-ylphenyl)-4-pyridinylmethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H23N3O
CAS Number:
Molecular Weight:
513.59
UNSPSC Code:
12352200
NACRES:
NA.23

assay

98%

form

solid

color

yellow

orbital energy

HOMO 5.75 eV 
LUMO 2.88 eV 

General description

The OLEDs using DCBPy, a benzoylpyridine-carbazole based fluorescence material as dopants emit blue light with EQEs of 24.0%, and with low efficiency roll-off at practical brightness level.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pachaiyappan Rajamalli et al.
Journal of the American Chemical Society, 138(2), 628-634 (2015-12-29)
Two benzoylpyridine-carbazole based fluorescence materials DCBPy and DTCBPy, bearing two carbazolyl and 4-(t-butyl)carbazolyl groups, respectively, at the meta and ortho carbons of the benzoyl ring, were synthesized. These molecules show very small ΔEST of 0.03 and 0.04 eV and transient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service