901304
Polyester bis-MPA dendron 16 NHBoc, 1 thiol (core)
generation 4
Synonym(s):
Bis-MPA dendron, PFd-G4-Thiol-NH-BOC
About This Item
description
16 NHBoc, 1 thiol (core)
form
solid
mol wt
generation 4
no. Surface Groups
16
color
white to off-white
functional group
thiol (core)
storage temp.
−20°C
General description
- Polymer architecture: Dendron
- End Group Functionality: Boc-protected amine
- Boc: tert-Butyloxycarbonyl
- Bis-MPA: 2,2-Bis(hydroxymethyl)propionic acid
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.
Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service