Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901300

Sigma-Aldrich

Polyester bis-MPA dendron 2 NHBoc, 1 thiol (core)

generation 1

Synonym(s):

Bis-MPA dendron, PFd-G1-Thiol-NH-BOC

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H40N2O10S
Molecular Weight:
536.64
UNSPSC Code:
12352105
NACRES:
NA.23

description

2 NHBoc, 1 thiol (core)

form

solid

mol wt

generation 1

no. Surface Groups

2

color

white to off-white

functional group

thiol (core)

storage temp.

−20°C

General description

  • Polymer architecture: Dendron
  • End Group Functionality: Boc-protected amine
  • Boc : tert-Butyloxycarbonyl
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid.

Application

Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers. Dendrons synthesized from bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. After Boc deprotection, amine-functionalized dendrons can be readily used in EDC or DCC coupling reactions with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications. Additionally, the thiol-core can be readily conjugated via a thiol-ene click reaction, Michael addition reactions, or disulfide exchange.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Articles

Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.

Dendrimers, polymer nanostructures, are synthesized for specific purposes by manipulating critical nanoscale design parameters (CNDPs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service