Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901265

Sigma-Aldrich

Gold/Silver cleaner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

form

liquid

General description

Removes oxides and tarnish from gold, silver, and precious metal alloy surfaces without leaving a residue. Gold/Silver cleaner is recommended for cleaning metal films immediately prior to further processing. Gold/Silver cleaner is an effective surface activator and adhesion promoter for gold and silver substrates prior to bonding processes.

Application

  • Substrates should be degreased first, if necessary.
  • Tank: Pyrex glass, fiberglass, polypropylene, etc.
  • Temperature: 20-50 °C.
  • Immersion Time: 30-60 seconds or as needed.
  • Rinse: water.
  • Use with agitation.

Features and Benefits

  • Rapid, economical cleaning.
  • Enhances fluxing, soldering, etching, plating, and coating.
  • No adverse electrical effects.
  • Mild pH.
  • Compatible with most metals.
  • Non-aggressive formulation.
  • Cyanide-free.

pictograms

Health hazardCorrosion

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 2 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service