Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901258

Sigma-Aldrich

1,1′-[4,8-Bis[4-(2-ethylhexyl)-3,5-difluorophenyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]bis[1,1,1-trimethylstannane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H58F4S2Sn2
CAS Number:
Molecular Weight:
964.47
UNSPSC Code:
12352101

assay

≥97%

form

powder or crystals

storage temp.

2-8°C

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangwu Li et al.
ACS applied materials & interfaces, 8(6), 3686-3692 (2015-12-10)
Two novel polymers PTFBDT-BZS and PTFBDT-BZO with 4-alkyl-3,5-difluorophenyl substituted benzodithiophene as the donor unit, benzothiadiazole or benzooxadiazole as the acceptor unit, and thiophene as the spacer have been synthesized and used as donor materials for polymer solar cells (PSCs). These

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service