Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901242

Sigma-Aldrich

5,10-Bis((5-octylthiophen-2-yl)dithieno[2,3-d:2′,3′-d′]benzo[1,2-b:4,5-b′]dithiophene-2,7-diyl)bis(trimethylstannane)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H58S6Sn2
CAS Number:
Molecular Weight:
1016.74
UNSPSC Code:
12352005
NACRES:
NA.23

assay

≥97%

form

powder or crystals

storage temp.

2-8°C

application

5,10-Bis((5-octylthiophen-2-yl)dithieno[2,3-d:2′,3′-d′]benzo[1,2-b:4,5-b′]dithiophene-2,7-diyl)bis(trimethylstannane) can be used in the synthesis of a wide bandgap polymer (PDBT-T1) for the fabrication of polymeric solar cells with an efficiency of 9.7%.[1]

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-junction organic solar cells based on a novel wide-bandgap polymer with efficiency of 9.7%.
Lijun Huo et al.
Advanced materials (Deerfield Beach, Fla.), 27(18), 2938-2944 (2015-04-03)

Articles

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service