Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901202

Sigma-Aldrich

1,1′-[4,8-Bis[5-(2-hexyldecyl)-2-thienyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]bis[1,1,1-trimethylstannane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H90S4Sn2
CAS Number:
Molecular Weight:
1128.99
UNSPSC Code:
12352005
NACRES:
NA.23

assay

97%

form

solid

storage temp.

2-8°C

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Side chain effect on poly(benzodithiophene-co-dithienobenzoquinoxaline) and their applications for polymer solar cells.
Fan J, et al.
Polymer, 82, 228-237 (2016)
Conjugated Side-Chain-Isolated D-A Copolymers Based on Benzo[1,2-b:4,5-b']dithiophene-alt-dithienylbenzotriazole: Synthesis and Photovoltaic Properties.
Min J, et al.
Chemistry of Materials, 24(16), 3247-3254 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service