Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901186

Sigma-Aldrich

Pyridyl disulfide ethyl methacrylate

≥98.0%, contains 150 ppm MEHQ as inhibitor

Synonym(s):

2-Pyridyl disulfide methacrylate, PDSEMA, PDSMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO2S2
CAS Number:
Molecular Weight:
255.36
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23

assay

≥98.0%

form

liquid

contains

150 ppm MEHQ as inhibitor

color

colorless to pale yellow

shipped in

wet ice

storage temp.

−20°C

General description

Pyridyl disulfide ethyl methacrylate is a methacylate-based monomer used in the synthesis of poly(pyridyl disulfide ethyl methacrylate) [p(PDSMA)] and other copolymers. p(PDSMA) contains a pendant, protected thiol that can be used for post-polymerization functionalization via thiol-disulfide exchange reactions. Additionally, reaction progress can be typically monitored by absorption spectroscopy as the spectroscopic signature of the released 2-pyridinethione (λ max = 375 nm) is distinctly different from the pyridyl disulfide functionality (λ max = 280 nm) in the polymer. Due to their versatility, these materials have been used in a wide variety of biomedical applications such as the synthesis of polymer-biomolecule conjugates and drug delivery applications.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive Copolymers Based on N-Vinyl Lactams with Pyridyl Disulfide Side Groups via RAFT Polymerization and Postmodification via Thiol-Disulfide Exchange Reaction.
Peng H, et al.
Macromolecules, 49(19), 7141-7154 (2016)
Xiaotian Ji et al.
Colloids and surfaces. B, Biointerfaces, 148, 41-48 (2016-11-05)
In this paper a novel method for the fabrication of hybrid nanogels based on thiol-disulfide exchange reaction is reported. Poly(oligo(ethylene glycol) monomethyl ether methacrylate-co-di(ethylene glycol) methyl ether methacrylate-co-2-(2-pyridyldisulfide) ethyl methacrylate) (POEGMA-co-PDEGMA-co-PDSMA) was synthesized by reversible addition-fragmentation chain transfer polymerization. Pyridyl
Simultaneous and Reversible Functionalization of Copolymers for Biological Applications.
Ghosh S, et al.
Macromolecules, 39(17), 5595-5597 (2006)
Judy Ventura et al.
Biomacromolecules, 16(10), 3161-3171 (2015-09-04)
Conjugation of biologically active proteins to polymeric materials is of great interest in the treatment of cancer and other diseases of protein deficiency. The conjugation of such biomacromolecules is challenging both due to their hydrophilicity and propensity to denature under
Nicholas M Matsumoto et al.
Polymer chemistry, 4(8), 2464-2469 (2014-04-25)
The covalent conjugation of bovine serum albumin (BSA) to disulfide cross-linked polymeric nanogels is reported. Polymeric nanogel precursors were synthesized via a reversible addition-fragmentation chain transfer (RAFT) random copolymerization of poly(ethylene glycol) methyl ether methacrylate (PEGMA) and pyridyl disulfide methacrylate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service