901142
SYLGARD® 186
10 g clip-pack
Synonym(s):
PDMS, Polydimethylsiloxane, Silicone elastomer
About This Item
Recommended Products
form
liquid
viscosity
123800 cP (part A or based)
66700 cP (mixed)
General description
Application
Potential Uses:
- General potting applications.
- Power supplies.
- Connectors.
- Sensors.
- Industrial controls.
- Transformers.
- Amplifiers.
- High voltage resistor packs.
- Relays.
- Adhesive/encapsulant for solar cells.
- Adhesive handling beam lead integrated circuits during processing.
Working time at 25 °C (pot life hours): 1.65 hr.
Heat cure time at 100 °C: 25 minutes.
Heat cure time at 125 °C: 15 minutes.
Heat cure time at 150 °C: 15 minutes.
Pot Life And Cure Rate - Cure reaction begins with the mixing process. Initially, cure is evidenced by a gradual increase in viscosity, followed by gelation and conversion to a solid elastomer. Pot life is defined as the time required for viscosity to double after Parts A and B (base and curing agent) are mixed and is highly temperature and application dependent. Please refer to the data table.
Useful Temperature Ranges - For most uses, silicone elastomers should be operational over a temperature range of -45 to 200°C (-49 to 392°F) for long periods of time. However, at both the low- and high temperature ends of the spectrum, behavior of the materials and performance in particular applications can become more complex and require additional considerations. For low temperature performance, thermal cycling to conditions such as -55°C (-67°F) may be possible, but performance should be verified for your parts or assemblies. Factors that may influence performance are configuration and stress sensitivity of components, cooling rates and hold times, and prior temperature history. At the high-temperature end, the durability of the cured silicone elastomer is time and temperature dependent. As expected, the higher the temperature, the shorter the time the material will remain useable.
Compatibility - Certain materials, chemicals, curing agents and plasticizers can inhibit the cure of addition cure adhesives. Most notable of these include: organotin and other organometallic compounds, silicone rubber containing organotin catalyst, sulfur, polysulfides, polysulfones or other sulfur containing materials, unsaturated hydrocarbon plasticizers, and some solder flux residues. If a substrate or material is questionable with respect to potentially causing inhibition of cure, it is recommended that a small scale compatibility test be run to ascertain suitability in a given application. The presence of liquid or uncured product at the interface between the questionable substrate and the cured gel indicates incompatibility and inhibition of cure.
Features and Benefits
- 10:1 mix ratio.
- High viscosity.
- RT and heat cure.
- High tear strength.
- UL tested.
Benefits:
- Rapid, versatile cure processing controlled by temperature.
- Can be considered for uses requiring UL requirements.
Preparation Note
Preparing Surfaces - In applications requiring adhesion, priming will be required for the silicone encapsulants. See the Primer Selection Guide for the correct primer to use with a given product. For best results, the primer should be applied in a very thin, uniform coating and then wiped off after application. After application, it should be thoroughly air dried prior to application of the silicone elastomer. Additional instructions for primer usage can be found in the information sheets specific to the individual primers.
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service