Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

901052

Sigma-Aldrich

Carbon nanotube array, multi-walled

vertically aligned on Si substrate, height 1.0 mm

Synonym(s):

CNT array, MWCNT array

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.23

description

Areal density: 1-5x109 cm-2
Form: 20 mmx20 mm Si substrate

assay

≥98% (carbon as CNT)

diam. × L

10-40 nm × 1.0 mm ± 0.1 mm

Looking for similar products? Visit Product Comparison Guide

General description

Multi-walled carbon nanotubes (MWCNT) are vertically aligned on a Si substrate. MWCNT is as long as millimetre scale. Crystal quality of CNT is examined by Raman scattering measurement. Intensity ratio of G-peak (1580 cm-1) and D-peak (1350 cm-1) represents quality of graphitization of carbon materials. G/D of this CNT is ~3, showing high crystal quality. The high crystallinity is supported by transmission electron microscopy.

Preparation Note

Chemical Vapor Deposition (CVD) method.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene nanoribbons (GNRs) are quasi-one-dimensional narrow strips of graphene comprised of sp2-hybridized carbon atoms arranged into hexagonal honeycomb lattice configurations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service