Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901024

Sigma-Aldrich

1-Propyl-3-methyl-imidazolium bis(trifluoromethylsulfonyl)imide Solarpur®

Synonym(s):

1-Methyl-3-propylimidazolium bis(trifluoromethanesulfonimide), PMIM NTF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13F6N3O4S2
CAS Number:
Molecular Weight:
405.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

assay

≥99.0% (HPLC)

form

liquid

SMILES string

O=S([N-]S(=O)(C(F)(F)F)=O)(C(F)(F)F)=O.CN1C=[N+](CCC)C=C1

InChI

1S/C7H13N2.C2F6NO4S2/c1-3-4-9-6-5-8(2)7-9;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H,3-4H2,1-2H3;/q+1;-1

InChI key

CDWUIWLQQDTHRA-UHFFFAOYSA-N

Application

Solarpur® electrolyte components for DSSC applications meet the highest purity standards regarding water and other impurities required for this technology.

Legal Information

Solarpur is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service