Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901023

Sigma-Aldrich

Poly(L-lactide-co-D,L-lactide)

70:30, viscosity 2.5 dL/g 

Synonym(s):

PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H8O4)m(C6H8O4)n
UNSPSC Code:
12162002

description

L-lactide: D,L-lactide: 70:30±5

form

chips

feed ratio

lactide:lactide 70:30

color

off-white to light yellow

viscosity

2.0-3.0 dL/g
2.5 dL/g

storage temp.

−20°C

General description

Poly(L-lactide-co-D,L-lactide) (PDLLA) is a biodegradable thermoplastic aliphatic polyester synthesized from both enantiomers (D and L) of lactic acid. Due to the ester linkages found in the polymer backbone, these copolymers are able to degrade via hydrolysis. Depending on the ratio of enantiomers used, different forms of PDLLA can be obtained. These differing forms yield materials with different degrees of crystallinity, thermal transitions, solubility, and rates of degradation.

Application

Poly(L-lactide-co-D,L-lactide) (PDLLA) is a biodegradable and biocompatible copolymer for use in biomedical research devices, such as implants, sutures, prosthetic devices, scaffolds for tissue engineering applications, micro and nanoparticles. PDLLA is also utilized in drug delivery research, such as in the synthesis and application of polymer-drug conjugates.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service