Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901003

Sigma-Aldrich

Poly(dioxanone-co-L-lactide)

90:10, viscosity 2.0 dL/g 

Synonym(s):

PDO-PLA, PDS-PLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H6O3)m(C6H8O4)n
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips

feed ratio

p-dioxanone:lactide 90:10±5

color

white to off-white

viscosity

1.5-2.5 dL/g
2.0 dL/g

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Poly(dioxanone-co-L-lactide) is a biodegradable copolymer synthesized from the ring-opening polymerization of p-dioxanone and lactide. Due to the ester linkages found in the polymer backbone, these polymers are able to degrade via hydrolysis. With different degrees of aliphatic character, the ratio of the two monomers affects the rate of hydrolysis and therefore the rate of degradation. Other factors that affect the degradation of these materials could include the end application or post-polymerization processing methods.

Application

Poly(dioxanone-co-L-lactide) is a biodegradable copolymer featuring the high strength of poly(dioxanone) and the flexibility of poly(L-lactide) for use in biomedical research applications such as biomedical devices, drug delivery systems, and tissue engineering constructs.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Main features of p-dioxanone, glycolide and lactide based copolymers

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service