Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900964

Sigma-Aldrich

DPTPCz

Synonym(s):

3-(4,6-Diphenyl-1,3,5-triazin-2-yl)-9-phenyl-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H22N4
CAS Number:
Molecular Weight:
474.55
UNSPSC Code:
12352116
NACRES:
NA.23

assay

≥97%

form

solid

Looking for similar products? Visit Product Comparison Guide

General description

  • A bipolar host material for highly efficient blue phosphorescent OLEDs.
  • TAPC:DPTPCz-based device showed a high EQE of 15.4%, which is the highest performance exciplex OLED up to date.[1]

application

DPTPCz, a carbazole based bipolar host material, can be used as an acceptor molecule for the fabrication of a photosensitizer. It can also be used in application such as phosphorescent organic light-emitting diodes (OLEDs).[2][3][4][5]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Ke Liu et al.
Advanced materials (Deerfield Beach, Fla.), 27(14), 2378-2383 (2015-02-26)
High-efficiency, thermally activated delayed-fluorescence organic light-emitting diodes based on exciplex emitters are demonstrated. The best device, based on a TAPC:DPTPCz emitter, shows a high external quantum efficiency of 15.4%. Strategies for predicting and designing efficient exciplex emitters are also provided.
Organic nanostructures of thermally activated delayed fluorescent emitters with enhanced intersystem crossing as novel metal-free photosensitizers
Zhang J, et al.
Chemical Communications (Cambridge, England), 52(79), 11744-11747 (2016)
Nearly 100% Triplet Harvesting in Conventional Fluorescent Dopant-Based Organic Light-Emitting Devices Through Energy Transfer from Exciplex
Liu X, et al.
Advanced Materials, 27(12), 2025-2030 (2015)
Highly efficient green and red phosphorescent OLEDs using novel bipolar blue fluorescent materials as hosts
Xiao J, et al.
Synthetic Metals, 193(12), 89-93 (2014)
Xiao-Ke Liu et al.
Advanced materials (Deerfield Beach, Fla.), 27(12), 2025-2030 (2015-02-14)
Nearly 100% triplet harvesting in conventional fluorophor-based organic light-emitting devices is realized through energy transfer from exciplex. The best C545T-doped device using the exciplex host exhibits a maximum current efficiency of 44.0 cd A(-1) , a maximum power efficiency of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service