Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900954

Sigma-Aldrich

PFN-DOF

Mw 5,000-10,000

Synonym(s):

Poly[(9,9-bis(3′-(N,N-dimethylamino)propyl)-2,7-fluorene)-alt-2,7-(9,9-dioctylfluorene)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C52H70N2)n
CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.23

form

solid

mol wt

Mw 5,000-10,000

solubility

chloroform: soluble
dichloromethane: soluble
toluene: soluble

λmax

378 nm in dichloromethane

fluorescence

λem 414 nm in dichloromethane (PL)

Looking for similar products? Visit Product Comparison Guide

Application

Phosphorescent host material. Polymer for use in electron transport layer in OPV and OLED applications.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Huang et al.
Journal of the American Chemical Society, 126(31), 9845-9853 (2004-08-05)
A series of aminoalkyl-substituted polyfluorene copolymers with benzothiadiazole (BTDZ) of different content were synthesized by Suzuki coupling reaction, and their quaternized ammonium polyelectrolyte derivatives were obtained through a postpolymerization treatment on the terminal amino groups. Copolymers are soluble in environmentally
Changfeng Han et al.
ACS applied materials & interfaces, 8(5), 3301-3307 (2016-01-13)
A highly efficient inverted polymer solar cell (PSC) has been successfully demonstrated by using a ZnO nanoparticle (NP) and poly[(9,9-bis(3'-(N,N-dimethylamino)propyl)-2,7-fluorene)-alt-2,7-(9,9-dioctyfluorene)] (PFN) bilayer structure as an effective electron collecting layer. This ZnO/PFN bilayer structure is designed to combine the advantages of
Novel Electroluminescent Conjugated Polyelectrolytes Based on Polyfluorene.
Huang F, et al.
Chemistry of Materials, 16(4), 708-716 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service