Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900939

Sigma-Aldrich

BPy-TP2

greener alternative

sublimed

Synonym(s):

2,7-Bis(2,2′-bipyridin-5-yl)triphenylene, 2,7-Di(2,2′-bipyridin-5-yl)triphenylene, 5-(7-[2,2′-Bipyridin]-5-yl-2-triphenylenyl)-2,2′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H24N4
CAS Number:
Molecular Weight:
536.62
UNSPSC Code:
12352103

assay

≥99% (sublimed)

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

334 °C

greener alternative category

General description

Electron transport or hole blocking material in OLEDs.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Electron transport organic materials have the chemical functionality to transport electrons and are used as electron transport layer in OLED devices. Click here for more information.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Color Tuning of Avobenzone Boron Difluoride as an Emitter to Achieve Full-Color Emission.
Mo HW, et al.
Advances in Functional Materials, 26(37), 6703-6710 (2016)
A solution-processable host material of 1,3-bis{3-[3-(9-carbazolyl)phenyl]-9-carbazolyl}benzene and its application in organic light-emitting diodes employing thermally activated delayed fluorescence.
Suzuki Y, et al.
Journal of Material Chemistry C, 3(8), 1700-1706 (2015)
Low driving voltage characteristics of triphenylene derivatives as electron transport materials in organic light-emitting diodes.
Togashi K, et al.
Journal of Materials Chemistry, 22(38), 20689-20695 (2012)

Articles

Explore the eco-friendly potential of organic thin film transistors (OTFTs) for detecting chemical analytes, identifying viruses, and assisting in health diagnostics. This mini-review highlights challenges of achieving sustainability, safety, and biodegradability of each component of an OTFT sensor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service