Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900884

Sigma-Aldrich

NIR-II dye

carboxylic acid functionalized

Synonym(s):

Near IR fluorescent dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100
NACRES:
NA.23

description

Mass extinction co-efficient: 3.5 Lg-1cm-1
viscous solid or liquid

form

liquid

λmax

780-810 nm

fluorescence

λem ~1050 nm, quantum yield ~2% (NIR emission)

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Stable solution in PBS. Miscible and stable in common buffers, serum and blood.
  • Store in dark at 4°C.
  • Suitable excitation wavelengths: 785 nm, 808 nm.
  • Emission: 900-1400 nm, peak ~ 1050 nm.
  • Quantum yield (QY) ~2% based on 0.5% QY of IR26 in 1,2 dichloroethane under 808 nm excitation. Quantum yield is ~4X of common Hipco single walled carbon nanotubes.
  • For in vivo imaging (e.g., vessels), inject ~100-150 μg in ~200 μL solution for a typical nude mouse but the dose can be varied.
  • Circulation half time of the dye is about 1 hour.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service