Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900830

Sigma-Aldrich

(2-{2-[2-(2-Methoxy-ethoxy)-ethoxy]-ethoxy}-ethyl)phosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17O6P
CAS Number:
Molecular Weight:
228.18
MDL number:
UNSPSC Code:
12352200

form

viscous liquid

Quality Level

availability

available only in USA

Application

This coupling agent is used to create antifouling surfaces.

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis Portilla et al.
Advanced materials (Deerfield Beach, Fla.), 27(39), 5950-5954 (2015-08-27)
TiO2 , Fe3 O4, AlOx , ITO (indium tin oxide), and CeO2 nanoparticles are tailored to exhibit excellent dispersability in deionized water and alcohols. The latter provides an ecofriendly solution for processing metal oxide nanoparticles at a neutral pH. Water-processed
Hanno Dietrich et al.
Advanced materials (Deerfield Beach, Fla.), 27(48), 8023-8027 (2015-11-03)
Tuning the electrostatics of ethylene-glycol-based self-assembled monolayers (SAMs) by doping with ions is shown. Molecular dynamics simulations unravel binding mechanisms and predict dipole strengths of the doped layers. Additionally, by applying such layers as dielectrics in organic thin-film transistors, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service