Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900822

Sigma-Aldrich

Polycaprolactone

viscosity 1.2 dL/g 

Synonym(s):

PCL, Poly[2-oxepanone], Purasorb PC 12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)nCH3
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

chunks

impurities

≤0.5% Residual monomer (GC)
≤0.5% Residual water content (colormetric titration)
≤100 ppm Tin (AAS)

inherent viscosity

1.0-1.3 dL/g

viscosity

1.2 dL/g

storage temp.

−20°C

InChI

1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2

InChI key

PAPBSGBWRJIAAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polycaprolactone (PCL) is a biodegradable, semicrystalline polyester for use in tissue engineering and drug delivery research applications. Due to the increased length of the aliphatic chain, polycaprolactone degrades significantly slower than other common biodegradable polymers, such as polylactide. PCL features a low melting point (55-60 °C), making it ideal for thermal processing and increasing its use in novel applications such as 3D bioprinting. In addition to its favorable thermal properties, PCL also features high solubility in organic solvent allowing for a multitude of other processing options. This product features low residual water, monomer, and catalyst (tin) making it an ideal choice for use in tissue engineering and 3D bioprinting research.

Application

Research applications of this material include:
  • Tissue engineering scaffolds.
  • 3D Bioprinting.
  • Drug delivery applications such as sustained release.

Analysis Note

Appearance: White to light tan granules.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Professor Mitsuhiro Ebara provides insights on several types of smart nanofiber mesh systems that have been explored for different drug delivery purposes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service