Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900786

Sigma-Aldrich

Silica

SBA-16, pellet, diam. × thickness 2 mm × 5 mm

Synonym(s):

Mesoporous slica, Mesostructured silica

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103

form

solid

diam. × thickness

2 mm × 5 mm

Looking for similar products? Visit Product Comparison Guide

Application

Unique physical properties such as high surface area and ordered pore structure make mesoporous silica useful for diverse applications extending from CO2 sequestration to catalysis and heavy metal removal from waste streams.[1][2][3] Our pelletized mesoporous materials are easy to handle and suitable for aforementioned applications especially in column format which is often required during continuous processing.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Gold Catalysts Supported on Mesoporous Silica Materials: Recent Developments.
Gutierrez LF, et al.
Catalysts, 1, 97-154 (2011)
Aoife M Burke et al.
Journal of hazardous materials, 164(1), 229-234 (2008-09-23)
Here we demonstrate aminopropyl and mercatopropyl functionalised and bi-functionalised large pore mesoporous silica spheres to extract various metal ions from aqueous solutions towards providing active sorbents for mitigation of metal ion pollution. Elemental analysis (EA) and FTIR techniques were used
Amrit Kumar et al.
Angewandte Chemie (International ed. in English), 54(48), 14372-14377 (2015-10-07)
Sequestration of CO2, either from gas mixtures or directly from air (direct air capture, DAC), could mitigate carbon emissions. Here five materials are investigated for their ability to adsorb CO2 directly from air and other gas mixtures. The sorbents studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service