Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900785

Sigma-Aldrich

Silica

mesoporous SBA-16, <150 μm particle size, pore size 5 nm, amine functionalized

Synonym(s):

Amine functionalized SBA-16, Amine functionalized mesoporous slica, Mesoporous slica

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352309

form

solid

particle size

<150 μm

pore size

5 nm pore size

functional group

amine

Looking for similar products? Visit Product Comparison Guide

Application

The unique physical properties such as high surface areas and ordered pore structure of mesoporous silica combined with the selectivity of amine and thiol functional groups make them useful for diverse applications. For example amine and thiol modified mesoporous silica finds applications in CO2 sequestration, heavy metal (such as Pb, Fe, Au etc.) removal from waste water or industrial waste streams.[1][2] Also, the ability of the amine and thiol modified mesoporous silica to bind the metallic nanoparticles such as Pt or Pd render them useful as novel catalytic support.[3] The functionalization of mesoporous silica with amine and thiol also makes them useful for entrapping biological entities and for investigations involving the study of the effect of quantum confinements of molecules/solvent within the ordered mesopores.[4][5]

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Gold Catalysts Supported on Mesoporous Silica Materials: Recent Developments.
Gutierrez LF, et al.
Catalysts, 1, 97-154 (2011)
Bob Grünberg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(22), 5689-5696 (2004-10-08)
The adsorption of water in two mesoporous silica materials with cylindrical pores of uniform diameter, MCM-41 and SBA-15, was studied by 1H MAS (MAS=magic angle spinning) and static solid-state NMR spectroscopy. All observed hydrogen atoms are either surface -SiOH groups
Chenghong Lei et al.
Journal of the American Chemical Society, 124(38), 11242-11243 (2002-09-19)
The enzyme organophosphorus hydrolase (OPH) was spontaneously entrapped in carboxylethyl- or aminopropyl-functionalized mesoporous silica with rigid, uniform open-pore geometry (30 nm). This approach yielded larger amounts of protein loading and much higher specific activity of the enzyme when compared to
Aoife M Burke et al.
Journal of hazardous materials, 164(1), 229-234 (2008-09-23)
Here we demonstrate aminopropyl and mercatopropyl functionalised and bi-functionalised large pore mesoporous silica spheres to extract various metal ions from aqueous solutions towards providing active sorbents for mitigation of metal ion pollution. Elemental analysis (EA) and FTIR techniques were used
Amrit Kumar et al.
Angewandte Chemie (International ed. in English), 54(48), 14372-14377 (2015-10-07)
Sequestration of CO2, either from gas mixtures or directly from air (direct air capture, DAC), could mitigate carbon emissions. Here five materials are investigated for their ability to adsorb CO2 directly from air and other gas mixtures. The sorbents studied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service