900723
Bovine tendon powder
neutral, type I collagen, suitable for biomedical research
About This Item
Recommended Products
description
Moisture content: Less than 12% (w/w)
form
powder
shelf life
24 mo. (after the date of manufacture)
impurities
<20 EU/g Endotoxin
color
white to off-white
pH
5.0-7.0
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Development of collagen-based tissues for skin replacement, bone substitutes and to generate artificial blood vessels and valves.
- Drug delivery systems such as sponges for burns/wounds, formulations with liposomes for sustained drug delivery, materials for transdermal drug delivery, and nanoparticles for gene delivery.
- As collagen scaffolds for research in cancer, electrophysiological protocols, neuron myelination, immunology and other cell culture applications.
Packaging
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Collagen-based delivery systems effectively deliver a range of viral and non-viral gene carriers by providing spatiotemporal control over delivery and reducing degradation rates and immune responses.
Protocols
Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service