Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900720

Sigma-Aldrich

PffBT4T-2OD

Synonym(s):

PCE-11, Poly[(5,6-difluoro-2,1,3-benzothiadiazol-4,7-diyl)-alt-(3,3’’’-di(2-octyldodecyl)-2,2’,5’,2’’,5’’,2’’’-quaterthiophen-5,5’’’-diyl)]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C62H88F2N2S5)n
CAS Number:
UNSPSC Code:
12352103

description

Band gap: 1.65 eV

form

powder

mol wt

Mw 100,000-150,000 by GPC

solubility

chlorinated solvents: soluble (at elevated temperature
solubility limit: <17 mg/ml in chlorobenzene + dichlorobenzene)

chlorobenzene: soluble
dichlorobenzene: soluble

λmax

400-750 nm

orbital energy

HOMO -5.34 eV 
LUMO -3.69 eV 

Application

Requires hot ink processing (i.e. coating with ink at temperature 100-110 °C to ensure good coatability).

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhang Liu et al.
Nature communications, 5, 5293-5293 (2014-11-11)
Although the field of polymer solar cell has seen much progress in device performance in the past few years, several limitations are holding back its further development. For instance, current high-efficiency (>9.0%) cells are restricted to material combinations that are
Zhihong Wu et al.
Journal of the American Chemical Society, 138(6), 2004-2013 (2016-01-23)
With the demonstration of small-area, single-junction polymer solar cells (PSCs) with power conversion efficiencies (PCEs) over the 10% performance milestone, the manufacturing of high-performance large-area PSC modules is becoming the most critical issue for commercial applications. However, materials and processes
Comparison of Two D-A Type Polymers with Each Being Fluorinated on D and A Unit for High Performance Solar Cells.
Jo JW, et al.
Advances in Functional Materials, 25(1), 120-125 (2015)

Articles

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service