Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900675

Sigma-Aldrich

Triaminetetraacetic acid-functionalized silica gel

≥99%

Synonym(s):

Si-TAAcOH, SiliaMetS® TAAcOH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.22

assay

≥99%

form

powder

shelf life

2 yr (expiration date period )

feature

endcapped: no

reaction suitability

reagent type: catalyst
reagent type: chelator

capacity

0.40 mmol/g loading(molecular loading)

solubility

all common solvents: soluble (aqueous and organic)

bulk density

635 g/L (typical tap density)

Application

Metal scavenger: Ca, Cu, Cs, Ir, Li, Mg, Ni, Os, Pd, and Sn.

Caution

For prolonged storage, recommended conditions are to store product in a cool place (<8 °C) under inert atmosphere (argon) when possible.

Legal Information

SiliaMetS is a registered trademark of Silicycle, Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service