Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900647

Sigma-Aldrich

1-Bromo-4-(1,2,2-triphenylethenyl)benzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H19Br
CAS Number:
Molecular Weight:
411.33
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

assay

98%

form

powder

InChI

1S/C26H19Br/c27-24-18-16-23(17-19-24)26(22-14-8-3-9-15-22)25(20-10-4-1-5-11-20)21-12-6-2-7-13-21/h1-19H

InChI key

MYJLJYSALGARCM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Changing the behavior of chromophores from aggregation-caused quenching to aggregation-induced emission: development of highly efficient light emitters in the solid state.
Wang Zhang Yuan et al.
Advanced materials (Deerfield Beach, Fla.), 22(19), 2159-2163 (2010-06-22)
Bhausaheb Dhokale et al.
The Journal of organic chemistry, 80(16), 8018-8025 (2015-07-21)
The tetraphenylethylene (TPE) substituted BODIPY 2a, and 2,3,3-triphenylacrylonitrile (TPAN) substituted BODIPYs 2b and 2c were designed and synthesized by the Pd-catalyzed Sonogashira cross-coupling reaction. Their photophysical and electrochemical properties were investigated. The BODIPY 2a exhibits strong D-A interaction with poor
New AIEgens containing tetraphenylethene and silole moieties: tunable intramolecular conjugation, aggregation-induced emission characteristics and good device performance.
Jie Y, et al.
Journal of Material Chemistry C, 3(11), 2624-2631 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service