Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900635

Sigma-Aldrich

Sodium 2-methylcyclopropylsulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO2S
Molecular Weight:
142.15
UNSPSC Code:
12352200

form

powder or crystals

reaction suitability

reaction type: C-C Bond Formation
reaction type: C-H Activation
reagent type: catalyst

Application

The following Baran sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed coupling of vinyl tosylates with arylsulfinate salts.
Reeves D C, et al.
Tetrahedron Letters, 50(24), 2870-2873 (2009)
Pd (II)-Catalyzed Direct Sulfonylation of Unactivated C (sp3)?H Bonds with Sodium Sulfinates.
Rao W H, et al.
Organic Letters, 17(14), 3552-3555 (2015)
Copper-Catalyzed Aerobic Decarboxylative Sulfonylation of Cinnamic Acids with Sodium Sulfinates: Stereospecific Synthesis of (E)-Alkenyl Sulfones.
Jiang Q, et al.
The Journal of Organic Chemistry, 79(16), 7372-7379 (2014)
Cu-Catalyzed Deoxygenative C2-Sulfonylation Reaction of Quinoline N-Oxides with Sodium Sulfinate.
Du B, et al.
Organic Letters, 18(16), 4144-4147 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service