Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900627

Sigma-Aldrich

15N2/N2(RG) Gas Mixture

ratio (1:80), ≥98 atom % 15N, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

isotopic purity

≥98 atom % 15N

assay

≥99% (CP)

form

gas

composition

ratio (1:80)

mass shift

M+2

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service