Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900498

Sigma-Aldrich

Methoxypolyethylene glycol pyrene

average Mn 2,000

Synonym(s):

mPEG-Pyrene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H17O(C2H4O)nCH3
UNSPSC Code:
12352200

form

powder

mol wt

average Mn 2,000

transition temp

flash point >230 °F
Tm 50-54 °C

storage temp.

2-8°C

Application

mPEG-Pyrene may be used for surface functionalization of carbon nanotubes and graphenes through π−π stacking between pyrene and aromatic rings on carbon nanotubes.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of hydrophobic interaction in the poly(methacrylic acid)/pyrene end-labeled poly(ethylene glycol) complex.
Oyama HT, et al.
Macromolecules, 20(8), 1839-1847 (1987)
Antonello Di Crescenzo et al.
Beilstein journal of nanotechnology, 5, 1675-1690 (2014-11-11)
Carbon nanotubes (CNTs) have been proposed and actively explored as multipurpose innovative nanoscaffolds for applications in fields such as material science, drug delivery and diagnostic applications. Their versatile physicochemical features are nonetheless limited by their scarce solubilization in both aqueous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service