Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900493

Sigma-Aldrich

Gold nano-urchins

80 nm, NHS ester functionalized, conjugation kit

Synonym(s):

NHS-activated gold nanourchins, Au nanoparticles, Gold nanoparticles, Urchins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141717
NACRES:
NA.23

form

nanourchin

shelf life

12 mo. at -20 °C (If stored unopened and as specified, are stable for at least 12 months)

storage condition

protect from light

particle size

80 nm

λmax

620 nm

functional group

NHS ester

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Drs. Fernandes and Baptista discuss gold nanoparticles synthesis and diagnostics regulations, focusing on lateral flow assays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service