900468
Gold nanoparticles
90 nm, maleimide functionalized, conjugation kit
Synonym(s):
Au nanoparticles
About This Item
form
nanoparticles
packaging
pkg of 1 ea
storage condition
protect from light
particle size
90 nm
λmax
564 nm
functional group
maleimide
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
1. Maleimide gold nanoparticles (lyophilized).
2. Ligand resuspension buffer.
3. Reaction buffer.
4. Quencher (lyophilized).
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Drs. Fernandes and Baptista discuss gold nanoparticles synthesis and diagnostics regulations, focusing on lateral flow assays.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service