Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900464

Sigma-Aldrich

Gold nanoparticles

50 nm, maleimide functionalized, conjugation kit

Synonym(s):

Au nanoparticles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141717
NACRES:
NA.23

form

nanoparticles

packaging

pkg of 1 ea

storage condition

protect from light

particle size

50 nm

λmax

535 nm

functional group

maleimide

Storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Biocompatibility, facile conjugation to biomolecules and unique optical properties arising from surface plasmon resonance render the nanoparticles of gold useful for variety of biomedical research such as bio-imaging, bio-sensing, photothermal hyperthermia, drug delivery etc. Our maleimide functionalized gold nanoparticles kits contain ready-to-use mixtures and have been optimized for high efficiency one-step conjugation of thiol-modified ligands such as oligonucleotides, antibodies, antibody fragments, proteins and peptides.
These kits contain the following components:
1. Maleimide gold nanoparticles (lyophilized).
2. Ligand resuspension buffer.
3. Reaction buffer.
4. Quencher (lyophilized).

Legal Information

Product of CytoDiagnostics, Inc.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Drs. Fernandes and Baptista discuss gold nanoparticles synthesis and diagnostics regulations, focusing on lateral flow assays.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service