Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900392

Sigma-Aldrich

Boc-hydroxylamine carboxylic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H26N2O6
Molecular Weight:
318.37
UNSPSC Code:
12352106
PubChem Substance ID:

assay

≥95%

form

solid or liquid

refractive index

n/D 1.4567

density

1.1544 g/mL

functional group

amine
carboxylic acid

Application

This hydroxylamine derivative readily reacts with potassium acyltrifluoroborates (KATs) to chemoselectively form amide bonds under aqueous conditions. Once incorporated into peptides, the Boc-hydroxylamine is stable to unprotected primary amines and conditions needed for ligation, deprotection, or Fmoc cleavage. The presence of the hydroxylamine on your peptide or protein enables the chemoselective conjugation of any of the several KAT analogues introduced collaboratively with the Jeff Bode Research Group.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Bode Group creates N-mesityl-NHC catalysts for enantioselective annulations and SnAP reagents for one-pot aldehyde conversion into N-heterocycles.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service