Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900360

Sigma-Aldrich

Ultroxa®: Poly(2-ethyl-2-oxazoline) amine terminated

average Mn 5,000, PDI ≤1.15

Synonym(s):

PEtOx-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C5H9NO)nNH2
UNSPSC Code:
12352200
NACRES:
NA.23

form

powder

mol wt

average Mn 5,000

PDI

≤1.15

storage temp.

2-8°C

Legal Information

Ultroxa is a registered trademark of University of Ghent

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The introduction of polymers into the biomedical field has opened new avenues in tissue engineering, implant design, biosensing, and drug delivery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service