Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900346

Sigma-Aldrich

3-Methyl-3-(2,3,5,6-tetrafluoro-4-nitrophenyl)-1,5-dioxaspiro[5.5]undecane-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13F4NO6
Molecular Weight:
391.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

powder

Storage temp.

2-8°C

SMILES string

FC1=C(F)C([N+]([O-])=O)=C(F)C(F)=C1C2(C)C(OC3(CCCCC3)OC2=O)=O

InChI

1S/C16H13F4NO6/c1-15(7-8(17)10(19)12(21(24)25)11(20)9(7)18)13(22)26-16(27-14(15)23)5-3-2-4-6-16/h2-6H2,1H3

Inchi Key

SZFIPJYAZVRMFV-UHFFFAOYSA-N

Application

This compound undergoes cycloreversion to form the fluoro(hetero)aryl ketene, which undergoes efficient coupling with nucleophiles and allows rapid incorporation of highly fluorinated α-fluoro(hetero)aryl acetic acid derivatives.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Organofluorine chemistry is an essential part of drug discovery programs as well as agrochemical programs and even plays a major role in materials chemistry. Despite the undeniable importance of fluorinated organic molecules, our ability to synthesize these substrates is lacking - though arguably it is better than that of Nature. Consequently, methods that allow facile access to fluorinated molecules are important especially when they provide unique access to fluorinated chemical space.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service