Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900334

Sigma-Aldrich

CdS/ZnS core-shell type quantum dots

oleic acid functionalized, fluorescence λem 450 nm, 5 mg/mL in toluene

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

dispersion

concentration

5 mg/mL in toluene

λmax

435 nm

fluorescence

λem 450 nm
λem 450 nm±10 nm, quantum yield >50%

functional group

oleic acid

Looking for similar products? Visit Product Comparison Guide

Application

Highly purified CdS/ZnS quantum dots with oleic acid surface ligand contain extremely low free organic ligands and impurities. They are well suited for display, solid state lighting, and solar cell applications. They can be used as electroluminescence emitter in quantum dots based LEDs or embedded in polymer matrix as the backlight of LCD display. CdSe/ZnS quantum dots in solid form can be dispersed in most non-polar organic solvents such as toluene, chloroform, and hexane.[1][2]

Caution

Do not freeze.

Legal Information

Product of Ocean Nanotech, LLC.

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F

flash_point_c

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hu W, et al.
Journal of the Electrochemical Society, 162, H747-H747 (2015)
Huaibin Shen et al.
ACS applied materials & interfaces, 5(22), 12011-12016 (2013-11-07)
In this paper, we use a simple device architecture based on solution-processed ZnO nanoparticles (NPs) as the electron injection/transport layer and bilayer structure of poly(ethylenedioxythiophene):polystyrene sulfonate (PEDOT:PSS)/poly[9,9-dioctylfluorene-co-N-[4-(3-methylpropyl)]-diphenylamine] (TFB) as the hole injection/transport layer to assess the effect of shell thickness

Articles

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service