Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900326

Sigma-Aldrich

Poly(p-dioxanone-co-glycolide-co-lactide

p-dioxanone:glycolide:lactide 90:5:5, viscosity 2.0 dL/g 

Synonym(s):

PDO-PGLA, PDS-PGLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C4H6O3)m(C4H4O4)n(C6H8O4)oCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules (chunks)

feed ratio

p-dioxanone:glycolide:lactide 90:5:5

viscosity

2.0 dL/g

transition temp

Tm 105-109 °C

storage temp.

−20°C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Main features of p-dioxanone, glycolide and lactide based copolymers

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service