Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900314

Sigma-Aldrich

Poly(caprolactone-co-glycolide)

caprolactone:glycolide 25:75, viscosity 1.6 dL/g 

Synonym(s):

PCL-PGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(C6H10O2)m(C4H4O4)nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips or granules

feed ratio

caprolactone:glycolide 25:75

viscosity

1.6 dL/g

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service