Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900283

Sigma-Aldrich

CdS/ZnS core-shell type quantum dots

oleic acid functionalized, fluorescence λem 425 nm, solid

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

form

powder
solid

mp

>380 °C

fluorescence

λem 425 nm
λem 425 nm±10 nm
, quantum yield >50%

functional group

oleic acid

Looking for similar products? Visit Product Comparison Guide

Application

Highly purified CdS/ZnS quantum dots with oleic acid surface ligand contain extremely low free organic ligands and impurities. They are well suited for display, solid state lighting, and solar cell applications. They can be used as electroluminescence emitter in quantum dots based LEDs or embedded in polymer matrix as the backlight of LCD display. CdSe/ZnS quantum dots in solid form can be dispersed in most non-polar organic solvents such as toluene, chloroform, and hexane.[1][2]

Caution

Do not freeze the material.

Legal Information

Product of Ocean Nanotech, LLC.

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Xu et al.
ACS applied materials & interfaces, 5(8), 2901-2907 (2013-04-05)
The excellent optical properties of quantum dots (QDs), such as high brightness, high photostability, continuous absorption, and narrow emission bandwidth, make them ideal as optical labels to develop QD-based immunohistofluorescence (IHF) imaging for multiplexing cancer biomarker detection on formalin-fixed and
Lily Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 5(2), 235-243 (2008-12-18)
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles
Zengyan Wei et al.
Soft matter, 8(26), 6871-6875 (2012-09-18)
Genetically-engineered collagen peptides were assembled into freestanding films when QDs are co-assembled as joints between collagen domains. These peptide based films show excellent mechanical properties with Young's modulus of ~20 GPa, much larger than most of multi-composite polymer films and

Articles

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service