Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900270

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

lyophilized, fluorescence λem 620 nm, solid

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
12352200
NACRES:
NA.23

form

lyophilized
powder
solid

λmax

605 nm

fluorescence

λem 620 nm
λem 620 nm±10 nm, quantum yield >50%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Reactive group: Carboxylic acid.

Application

CdS/ZnS or CdSe/ZnS quantum dots (QDs) with carboxylic acid in lyophilized solid form can be readily reconstituted in DI water or DMSO to any desired concentration and are very stable in most buffer solutions with a pH range of 5-10. These QDs possess a zeta potential in the -30 mV to -50 mV range and can easily be reacted with amine containing molecules in DMSO or water. These QDs find applications in bioimaging and biosensing.

Legal Information

Product of Ocean Nanotech, LLC.

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zengyan Wei et al.
Soft matter, 8(26), 6871-6875 (2012-09-18)
Genetically-engineered collagen peptides were assembled into freestanding films when QDs are co-assembled as joints between collagen domains. These peptide based films show excellent mechanical properties with Young's modulus of ~20 GPa, much larger than most of multi-composite polymer films and
Hong Xu et al.
ACS applied materials & interfaces, 5(8), 2901-2907 (2013-04-05)
The excellent optical properties of quantum dots (QDs), such as high brightness, high photostability, continuous absorption, and narrow emission bandwidth, make them ideal as optical labels to develop QD-based immunohistofluorescence (IHF) imaging for multiplexing cancer biomarker detection on formalin-fixed and
Lily Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 5(2), 235-243 (2008-12-18)
Epidermal growth factor receptor (EGFR) targeted nanoparticle are developed by conjugating a single-chain anti-EGFR antibody (ScFvEGFR) to surface functionalized quantum dots (QDs) or magnetic iron oxide (IO) nanoparticles. The results show that ScFvEGFR can be successfully conjugated to the nanoparticles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service