Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900246

Sigma-Aldrich

CdSe/ZnS core-shell type quantum dots

PEG functionalized, fluorescence λem 580 nm, 4 μM in H2O

Synonym(s):

Fluorescent nanocrystals, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSe/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

form

dispersion

Quality Level

concentration

4 μM in H2O

fluorescence

λem 580 nm
λem 580 nm±10 nm, quantum yield >50%

functional group

PEG

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Reactive group: None.

Application

The CdS/ZnS or CdSe/ZnS quantum dots functionalized with PEG exhibit low non-specific binding which makes them suitable for a variety of in vivo and in vitro biomedical applications.[1]

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiangqiang Liu et al.
International journal of nanomedicine, 10, 7073-7088 (2015-11-26)
The growing potential of quantum dots (QDs) in biomedical applications has provoked the urgent need to thoroughly address their interaction with biological systems. However, only limited studies have been performed to explore the effects of surface charge on the biological

Articles

Cadmium-free quantum dots for bioimaging offer design and synthesis strategies, enhancing bioimaging and sensing applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service